COMH4211B9B3 Combi-Blocks, Inc.

2-Pyridinamine, 3-[(2,6-difluorophenyl)methoxy]-

  • MDL number MFCD29086868
  • Cas Number : 151411-13-1
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5838
Availability details for
Quantity
  • Availability
  • Add to Cart