COMH4211B9B5 Combi-Blocks, Inc.

4-Bromo-2-isobutoxyphenylamine

  • MDL number MFCD21193996
  • Cas Number : 1490694-65-9
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5846

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5846
Availability details for
Quantity
  • Availability
  • Add to Cart