COMH4211B9B8 Combi-Blocks, Inc.

5-Bromo-2-(2-methoxy-ethoxy)-benzoic acid methyl ester

  • MDL number MFCD17292514
  • Cas Number : 75230-40-9
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5853
Availability details for
Quantity
  • Availability
  • Add to Cart