COMH4211B9B9 Combi-Blocks, Inc.

6-(4-(Trifluoromethyl)phenyl)pyridazin-3-amine

  • MDL number MFCD14707058
  • Cas Number : 1215073-83-8
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5856
Availability details for
Quantity
  • Availability
  • Add to Cart