COMH4211B9BB Combi-Blocks, Inc.

4-Bromo-2-fluoro-5-(trifluoromethoxy)aniline

  • MDL number MFCD22047211
  • Cas Number : 116367-67-0
  • Molecular Weight : 274.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5860

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5860
Availability details for
Quantity
  • Availability
  • Add to Cart