COMH4211B9BC Combi-Blocks, Inc.

L-Phenylalanine, n-[2-(2-naphthalenyl)acetyl]-

  • MDL number MFCD31381643
  • Cas Number : 934395-57-0
  • Molecular Weight : 333.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5867
Availability details for
Quantity
  • Availability
  • Add to Cart