COMH4211B9BE Combi-Blocks, Inc.

1-(3-Bromo-4-propoxy-phenyl)-ethanone

  • MDL number MFCD17247612
  • Cas Number : 35310-77-1
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5869

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5869
Availability details for
Quantity
  • Availability
  • Add to Cart