COMH4211B9C0 Combi-Blocks, Inc.

Methyl 2-(acetylthio)-4-bromobutanoate

  • MDL number MFCD31561576
  • Cas Number : 1193111-33-9
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5871
Availability details for
Quantity
  • Availability
  • Add to Cart