COMH4211B9C2 Combi-Blocks, Inc.

2-(5-Phenyl-1,3-oxazol-2-yl)acetonitrile

  • MDL number MFCD13188033
  • Cas Number : 343774-44-7
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5874
Availability details for
Quantity
  • Availability
  • Add to Cart