COMH4211B9C6 Combi-Blocks, Inc.

2-Fluoro-4-[2-(tetrahydro-pyran-2-yloxy)-ethoxy]-phenylamine

  • MDL number MFCD31614002
  • Cas Number : 873453-88-4
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5883
Availability details for
Quantity
  • Availability
  • Add to Cart