COMH4211B9C7 Combi-Blocks, Inc.

2-(4-Cyclopropylmethoxy-pyrazol-1-yl)-2-methyl-propionic acid

  • MDL number MFCD31579343
  • Cas Number : 2270905-34-3
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5886
Availability details for
Quantity
  • Availability
  • Add to Cart