COMH4211B9C8 Combi-Blocks, Inc.

3-Bromo-5-(trifluoromethyl)benzamide

  • MDL number MFCD27980429
  • Cas Number : 1007578-83-7
  • Molecular Weight : 268.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5889
Availability details for
Quantity
  • Availability
  • Add to Cart