COMH4211B9C9 Combi-Blocks, Inc.

5-Bromo-3-[(4-methoxybenzyl)thio]-2-pyrazinamine

  • MDL number MFCD30720696
  • Cas Number : 2096987-62-9
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5895
Availability details for
Quantity
  • Availability
  • Add to Cart