COMH4211B9CB Combi-Blocks, Inc.

3-Bromo-5-cyclopropanesulfonylamino-benzoic acid

  • MDL number MFCD25629357
  • Cas Number : 1540171-83-2
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5897
Availability details for
Quantity
  • Availability
  • Add to Cart