COMH4211B9CC Combi-Blocks, Inc.

(2-(2-[2-(6-Amino-hexanoylamino)-ethoxy]-ethoxy)-ethyl)-carbamic acid tert-butyl ester

  • MDL number MFCD31579398
  • Cas Number : 2270905-53-6
  • Molecular Weight : 361.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5898
Availability details for
Quantity
  • Availability
  • Add to Cart