COMH4211B9D1 Combi-Blocks, Inc.

4-(1-Ethyl-propoxy)-benzylamine

  • MDL number MFCD23832722
  • Cas Number : 1505292-74-9
  • Molecular Weight : 193.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5909

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5909
Availability details for
Quantity
  • Availability
  • Add to Cart