COMH4211B9D9 Combi-Blocks, Inc.

3-Bromo-4-fluoro-2,n-dimethyl-benzamide

  • MDL number MFCD31579363
  • Cas Number : 2270905-22-9
  • Molecular Weight : 246.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5931
Availability details for
Quantity
  • Availability
  • Add to Cart