COMH4211B9DA Combi-Blocks, Inc.

2-(Cyclobutylamino)-5-fluorobenzoic acid

  • MDL number MFCD31613846
  • Cas Number : 1593887-47-8
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5932

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5932
Availability details for
Quantity
  • Availability
  • Add to Cart