COMH4211B9DD Combi-Blocks, Inc.

Toluene-4-sulfonic acid 2-[2-(2-allyloxy-ethoxy)-ethoxy]-ethyl ester

  • MDL number MFCD31381596
  • Cas Number : 84183-97-1
  • Molecular Weight : 344.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5940
Availability details for
Quantity
  • Availability
  • Add to Cart