COMH4211B9DF Combi-Blocks, Inc.

2-(4-(Trifluoromethyl)phenyl)thiazole

  • MDL number MFCD12033578
  • Cas Number : 119514-24-8
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5948
Availability details for
Quantity
  • Availability
  • Add to Cart