COMH4211B9E1 Combi-Blocks, Inc.

3-Bromo-n-(2,2,2-trifluoroethyl)benzamide

  • MDL number MFCD09945038
  • Cas Number : 26930-20-1
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5950

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5950
Availability details for
Quantity
  • Availability
  • Add to Cart