COMH4211B9E5 Combi-Blocks, Inc.

4-[2-Fluoro-4-(2,2,2-trifluoro-ethoxy)-phenyl]-1,2,3,6-tetrahydro-pyridine hydrochloride

  • MDL number MFCD31583388
  • Cas Number : 2270913-18-1
  • Molecular Weight : 311.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5964
Availability details for
Quantity
  • Availability
  • Add to Cart