COMH4211B9E6 Combi-Blocks, Inc.

4-(2-Bromophenoxy)benzoic acid

  • MDL number MFCD11156573
  • Cas Number : 86607-80-9
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5967
Availability details for
Quantity
  • Availability
  • Add to Cart