COMH4211B9EC Combi-Blocks, Inc.

Z-Ile-gly-oh

  • MDL number MFCD00020454
  • Cas Number : 13254-04-1
  • Molecular Weight : 322.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5983

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5983
Availability details for
Quantity
  • Availability
  • Add to Cart