COMH4211B9ED Combi-Blocks, Inc.

4-(Cyclohexylmethyl)aniline

  • MDL number MFCD11610553
  • Cas Number : 145071-73-4
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5986
Availability details for
Quantity
  • Availability
  • Add to Cart