COMH4211B9EE Combi-Blocks, Inc.

5-Benzyloxy-2-isopropoxy-benzoic acid

  • MDL number MFCD31618902
  • Cas Number : 176674-45-6
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5990

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5990
Availability details for
Quantity
  • Availability
  • Add to Cart