COMH4211B9F3 Combi-Blocks, Inc.

2-Chloro-n-pyridin-3-ylisonicotinamide

  • MDL number MFCD01934954
  • Cas Number : 680217-96-3
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5998
Availability details for
Quantity
  • Availability
  • Add to Cart