COMH4211B9F4 Combi-Blocks, Inc.

2-Amino-4-phenylbutan-1-ol

  • MDL number MFCD09744981
  • Cas Number : 160886-95-3
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6002
Availability details for
Quantity
  • Availability
  • Add to Cart