COMH4211B9F5 Combi-Blocks, Inc.

5-Iodomethyl-2-methyl-pyrimidine

  • MDL number MFCD30188003
  • Cas Number : 2090297-94-0
  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6004
Availability details for
Quantity
  • Availability
  • Add to Cart