COMH4211B9F6 Combi-Blocks, Inc.

3-Bromo-5-cyclopropylmethoxy-n-methoxy-n-methyl-benzamide

  • MDL number MFCD31381608
  • Cas Number : 1180016-75-4
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6005
Availability details for
Quantity
  • Availability
  • Add to Cart