COMH4211B9F7 Combi-Blocks, Inc.

3-(4-Fluoro-benzenesulfonyl)-propylamine hydrochloride

  • MDL number MFCD15201833
  • Cas Number : 1158290-01-7
  • Molecular Weight : 253.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6007
Availability details for
Quantity
  • Availability
  • Add to Cart