COMH4211B9F9 Combi-Blocks, Inc.

3-Fluoro-2-(2-methoxy-ethoxy)-benzylamine hydrochloride

  • MDL number MFCD31557797
  • Cas Number : 2205505-30-0
  • Molecular Weight : 235.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6011
Availability details for
Quantity
  • Availability
  • Add to Cart