COMH4211B9FA Combi-Blocks, Inc.

3-Nitro-2,4,6-trifluorobenzoic acid

  • MDL number MFCD28801814
  • Cas Number : 1803842-19-4
  • Molecular Weight : 221.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6015

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6015
Availability details for
Quantity
  • Availability
  • Add to Cart