COMH4211B9FB Combi-Blocks, Inc.

1-(4-Isopropoxyphenyl)propan-2-one

  • MDL number MFCD00461574
  • Cas Number : 84023-37-0
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6016

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6016
Availability details for
Quantity
  • Availability
  • Add to Cart