COMH4211B9FE Combi-Blocks, Inc.

5-Iodopentanenitrile

  • MDL number MFCD20415271
  • Cas Number : 6727-75-9
  • Molecular Weight : 209.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6019
Availability details for
Quantity
  • Availability
  • Add to Cart