COMH4211BA02 Combi-Blocks, Inc.

4-[4-(2,4-Dioxothiazolidin-5-ylidenemethyl)phenoxy]butyric acid

  • MDL number MFCD31579418
  • Cas Number : 503827-51-8
  • Molecular Weight : 307.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6023
Availability details for
Quantity
  • Availability
  • Add to Cart