COMH4211BA03 Combi-Blocks, Inc.

4-Isoxazolemethanol, 3-(2,3-difluorophenyl)-5-methyl-

  • MDL number MFCD22370401
  • Cas Number : 1159602-37-5
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6024
Availability details for
Quantity
  • Availability
  • Add to Cart