COMH4211BA05 Combi-Blocks, Inc.

5-(3-Methylbutyl)-1h-pyrazole-3-carboxylic acid

  • MDL number MFCD20657668
  • Cas Number : 856256-86-5
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-isopentyl-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C9H14N2O2/c1-6(2)3-4-7-5-8(9(12)13)11-10-7/h5-6H,3-4H2,1-2H3,(H,10,11)(H,12,13)
Inchi Key CTBFZIBBKPFHDW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6026
Availability details for
Quantity
  • Availability
  • Add to Cart