COMH4211BA07 Combi-Blocks, Inc.

4-(4-Isopropoxy-phenyl)-butan-2-one

  • MDL number MFCD09950459
  • Cas Number : 1016851-63-0
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6030
Availability details for
Quantity
  • Availability
  • Add to Cart