COMH4211BA09 Combi-Blocks, Inc.

2-(4-Bromo-phenyl)-5-methyl-imidazo[5,1-b][1,2,4]oxadiazole

  • MDL number MFCD31579356
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6034
Availability details for
Quantity
  • Availability
  • Add to Cart