COMH4211BA0A Combi-Blocks, Inc.

4-Bromo-3-(2,2,2-trifluoroethoxy)benzoic acid

  • MDL number MFCD30536266
  • Cas Number : 1226966-58-0
  • Molecular Weight : 299.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6035
Availability details for
Quantity
  • Availability
  • Add to Cart