COMH4211BA0B Combi-Blocks, Inc.

(2S)-Glycidyl indol-4-yl ether

  • MDL number MFCD31614110
  • Cas Number : 70260-86-5
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6038
Availability details for
Quantity
  • Availability
  • Add to Cart