COMH4211BA0C Combi-Blocks, Inc.

5-Fluoro-2-isobutoxyaniline

  • MDL number MFCD11135818
  • Cas Number : 640768-04-3
  • Molecular Weight : 183.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6039

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6039
Availability details for
Quantity
  • Availability
  • Add to Cart