COMH4211BA0D Combi-Blocks, Inc.

4-Benzyloxy-2-nitro-benzoic acid

  • MDL number MFCD31583113
  • Cas Number : 1775656-80-8
  • Molecular Weight : 273.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6041
Availability details for
Quantity
  • Availability
  • Add to Cart