COMH4211BA0E Combi-Blocks, Inc.

4-(2-Fluoro-4-isobutoxy-phenyl)-piperidine hydrochloride

  • MDL number MFCD31583392
  • Cas Number : 2270909-43-6
  • Molecular Weight : 287.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6044
Availability details for
Quantity
  • Availability
  • Add to Cart