COMH4211BA10 Combi-Blocks, Inc.

N-Cyclopropyl-5-hydroxy-2-isopropoxy-benzamide

  • MDL number MFCD31618909
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6047

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6047
Availability details for
Quantity
  • Availability
  • Add to Cart