COMH4211BA11 Combi-Blocks, Inc.

2-Bromo-4-(cyclopropylmethoxy)-1-fluorobenzene

  • MDL number MFCD26024414
  • Cas Number : 1369814-15-2
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-(cyclopropylmethoxy)-1-fluorobenzene
Inchi Code 1S/C10H10BrFO/c11-9-5-8(3-4-10(9)12)13-6-7-1-2-7/h3-5,7H,1-2,6H2
Inchi Key LSSAQUSKFUYLPB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6048
Availability details for
Quantity
  • Availability
  • Add to Cart