COMH4211BA12 Combi-Blocks, Inc.

2-Iodo-3-cyclopropylmethoxypyridine

  • MDL number MFCD25960822
  • Cas Number : 205594-62-3
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6050
Availability details for
Quantity
  • Availability
  • Add to Cart