COMH4211BA13 Combi-Blocks, Inc.

4-(2-Bromo-5-nitro-benzyl)-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31614011
  • Cas Number : 2270907-32-7
  • Molecular Weight : 400.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6051
Availability details for
Quantity
  • Availability
  • Add to Cart