COMH4211BA14 Combi-Blocks, Inc.

4-(2-Fluoro-4-hydroxy-phenyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583380
  • Cas Number : 1509045-13-9
  • Molecular Weight : 295.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6053
Availability details for
Quantity
  • Availability
  • Add to Cart